The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000158.3 | NP_000149.3 | 14 - 15 | 2221 | 59 |
GenBank mRNA | AB209731.1 | - | 14 - 15 | 2111 | 59 |
AK125918.1 | - | 14 - 15 | 2584 | 59 | |
AK297939.1 | - | 12 - 13 | 1654 | 59 | |
AK300620.1 | - | 14 - 15 | 1868 | 59 | |
AK312245.1 | - | 14 - 15 | 2069 | 59 | |
BC012098.1 | - | 14 - 15 | 2150 | 59 | |
L07956.1 | - | 14 - 15 | 2028 | 59 |