Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAD2
Entrez Gene ID
2572
Gene Name
glutamate decarboxylase 2
Gene Aliases
GAD65
Chromosome Location
Chr.10: 26216307 - 26304562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000818.2 NP_000809.1 10 - 11 1597 134
NM_001134366.1 NP_001127838.1 10 - 11 1597 134
GenBank mRNA AJ251501.1 - 7 - 8 597 134
BC126327.1 - 10 - 11 1119 134
BC126329.1 - 10 - 11 1119 134
M74826.1 - 10 - 11 1235 134
M81882.1 - 10 - 11 1146 134
X69936.1 - 10 - 11 1139 134

View Full Product Details