The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001196.3 | NP_001187.1 | 2 - 3 | 190 | 72 |
NM_001244567.1 | NP_001231496.1 | 2 - 3 | 101 | 72 | |
NM_197966.2 | NP_932070.1 | 2 - 3 | 508 | 72 | |
XM_017028906.1 | XP_016884395.1 | 2 - 3 | 166 | 72 | |
GenBank mRNA | AF042083.1 | - | 2 - 3 | 157 | 72 |
AF087891.1 | - | 2 - 3 | 155 | 72 | |
AF250233.1 | - | 2 - 3 | 449 | 72 | |
AK094795.1 | - | 2 - 3 | 449 | 72 | |
AK313729.1 | - | 2 - 3 | 145 | 72 | |
AY005151.1 | - | 2 - 3 | 107 | 72 | |
BC009197.2 | - | 2 - 3 | 152 | 72 | |
BC022072.1 | - | 2 - 3 | 190 | 72 | |
BC033634.1 | - | 2 - 3 | 159 | 72 | |
BC036364.1 | - | 2 - 3 | 483 | 72 | |
BM561391.1 | - | 2 - 3 | 201 | 72 | |
BQ068464.1 | - | 2 - 3 | 168 | 72 | |
CR456389.1 | - | 1 - 2 | 59 | 72 | |
D89803.1 | - | 1 - 2 | 90 | 72 | |
DA274603.1 | - | 2 - 3 | 508 | 72 | |
EU678292.1 | - | 2 - 3 | 107 | 72 | |
EU678294.1 | - | 2 - 3 | 101 | 72 |