The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256283.1 | NP_001243212.1 | 2 - 3 | 592 | 88 |
NM_001957.3 | NP_001948.1 | 3 - 4 | 1082 | 88 | |
GenBank mRNA | AK304451.1 | - | 2 - 3 | 364 | 88 |
AK312812.1 | - | 3 - 4 | 902 | 88 | |
AK315931.1 | - | 2 - 3 | 577 | 88 | |
AY275462.1 | - | 2 - 3 | 552 | 88 | |
BC022511.1 | - | 3 - 4 | 902 | 88 | |
D90348.1 | - | 3 - 4 | 1032 | 88 | |
L06622.1 | - | 2 - 3 | 558 | 88 | |
S45956.1 | - | 3 - 4 | 773 | 88 | |
S57498.1 | - | 3 - 4 | 1036 | 88 | |
S63938.1 | - | 2 - 3 | 625 | 88 | |
S67127.1 | - | 2 - 3 | 689 | 88 | |
S81539.1 | - | 2 - 3 | 552 | 88 | |
X61950.1 | - | 3 - 4 | 1036 | 88 |