Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
T
Entrez Gene ID
6862
Gene Name
T brachyury transcription factor
Gene Aliases
SAVA, TFT
Chromosome Location
Chr.6: 166157644 - 166168669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270484.1 NP_001257413.1 4 - 5 1033 83
NM_003181.3 NP_003172.1 4 - 5 1126 83
XM_011536080.2 XP_011534382.1 3 - 4 893 83
GenBank mRNA AJ001699.1 - 3 - 4 766 83
BC039877.1 - 3 - 4 893 83
BC098425.1 - 4 - 5 1006 83

View Full Product Details