The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270484.1 | NP_001257413.1 | 5 - 6 | 1095 | 107 |
NM_003181.3 | NP_003172.1 | 5 - 6 | 1188 | 107 | |
XM_011536080.2 | XP_011534382.1 | 4 - 5 | 955 | 107 | |
GenBank mRNA | AJ001699.1 | - | 4 - 5 | 828 | 107 |
BC039877.1 | - | 4 - 5 | 955 | 107 | |
BC098425.1 | - | 5 - 6 | 1068 | 107 |