Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 6 - 7 1242 63
NM_001322988.1 NP_001309917.1 6 - 7 1242 63
NM_001322989.1 NP_001309918.1 6 - 7 1242 63
NM_001322990.1 NP_001309919.1 6 - 7 1242 63
NM_001322991.1 NP_001309920.1 6 - 7 1242 63
NM_001322992.1 NP_001309921.1 6 - 7 1242 63
NM_003947.5 NP_003938.1 6 - 7 1242 63
XM_006713810.3 XP_006713873.1 6 - 7 1487 63
XM_006713811.3 XP_006713874.1 6 - 7 1487 63
XM_006713812.3 XP_006713875.1 6 - 7 1487 63
XM_006713813.3 XP_006713876.1 6 - 7 1487 63
XM_006713814.3 XP_006713877.1 6 - 7 1487 63
XM_006713815.3 XP_006713878.1 6 - 7 1490 63
XM_011513279.2 XP_011511581.1 6 - 7 1487 63
XM_011513280.2 XP_011511582.1 6 - 7 1488 63
XM_011513281.2 XP_011511583.1 6 - 7 24136 63
XM_011513283.2 XP_011511585.1 6 - 7 1484 63
XM_011513285.2 XP_011511587.1 6 - 7 1483 63
XM_017007429.1 XP_016862918.1 6 - 7 1487 63
XM_017007430.1 XP_016862919.1 6 - 7 1487 63
XM_017007431.1 XP_016862920.1 6 - 7 1483 63
XM_017007432.1 XP_016862921.1 6 - 7 1483 63
XM_017007433.1 XP_016862922.1 6 - 7 1490 63
XM_017007434.1 XP_016862923.1 6 - 7 1490 63
XM_017007435.1 XP_016862924.1 6 - 7 1490 63
GenBank mRNA AK314126.1 - 6 - 7 1242 63
BQ345377.1 - 1 - 2 63
U94190.1 - 6 - 7 1185 63

View Full Product Details