Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72229432-72229693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850225 c.772G>A p.E258K Chr.11: 72229681-72229681 on Build GRCh38
6318626 c.719G>T p.S240I Chr.11: 72229524-72229524 on Build GRCh38
1605186 c.683G>T p.G228V Chr.11: 72229488-72229488 on Build GRCh38
6451242 c.747G>A p.Q249Q Chr.11: 72229552-72229552 on Build GRCh38
4507792 c.756C>T p.N252N Chr.11: 72229665-72229665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776498366 Chr.11: 72229594-72229594 on Build GRCh38
rs760513714 Chr.11: 72229527-72229527 on Build GRCh38
rs750527388 Chr.11: 72229646-72229646 on Build GRCh38
rs753707673 Chr.11: 72229532-72229532 on Build GRCh38
rs775043885 Chr.11: 72229655-72229655 on Build GRCh38
rs773348902 Chr.11: 72229526-72229526 on Build GRCh38
rs761235393 Chr.11: 72229496-72229496 on Build GRCh38
rs375761722 Chr.11: 72229635-72229635 on Build GRCh38
rs756452267 Chr.11: 72229559-72229559 on Build GRCh38
rs377498321 Chr.11: 72229585-72229585 on Build GRCh38
rs756574741 Chr.11: 72229651-72229651 on Build GRCh38
rs367992744 Chr.11: 72229588-72229588 on Build GRCh38
rs747802572 Chr.11: 72229671-72229671 on Build GRCh38
rs771023319 Chr.11: 72229593-72229593 on Build GRCh38
rs778596194 Chr.11: 72229664-72229664 on Build GRCh38
rs766325854 Chr.11: 72229528-72229528 on Build GRCh38
rs763192141 Chr.11: 72229537-72229537 on Build GRCh38
rs773871418 Chr.11: 72229488-72229488 on Build GRCh38
rs757335516 Chr.11: 72229543-72229543 on Build GRCh38
rs201713633 Chr.11: 72229659-72229659 on Build GRCh38
rs749022309 Chr.11: 72229462-72229462 on Build GRCh38
rs774695734 Chr.11: 72229544-72229544 on Build GRCh38
rs771989660 Chr.11: 72229524-72229524 on Build GRCh38
rs374049262 Chr.11: 72229660-72229660 on Build GRCh38
rs768216134 Chr.11: 72229485-72229485 on Build GRCh38
rs759259120 Chr.11: 72229595-72229595 on Build GRCh38
rs770890568 Chr.11: 72229507-72229507 on Build GRCh38
rs45506896 Chr.11: 72229589-72229589 on Build GRCh38
rs749646417 Chr.11: 72229578-72229578 on Build GRCh38
rs780351983 Chr.11: 72229576-72229576 on Build GRCh38
rs867029347 Chr.11: 72229652-72229652 on Build GRCh38
rs915157319 Chr.11: 72229639-72229639 on Build GRCh38
rs764222606 Chr.11: 72229539-72229539 on Build GRCh38
rs774501149 Chr.11: 72229624-72229624 on Build GRCh38
rs573385475 Chr.11: 72229584-72229584 on Build GRCh38
rs766877002 Chr.11: 72229658-72229658 on Build GRCh38
rs1052407023 Chr.11: 72229592-72229592 on Build GRCh38
rs750970470 Chr.11: 72229549-72229549 on Build GRCh38
rs891799988 Chr.11: 72229654-72229654 on Build GRCh38
rs554820987 Chr.11: 72229540-72229540 on Build GRCh38
rs12795135 Chr.11: 72229605-72229605 on Build GRCh38
rs878853120 Chr.11: 72229558-72229558 on Build GRCh38

View Full Product Details