Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUS3L
Entrez Gene ID
56931
Gene Name
dihydrouridine synthase 3 like
Gene Aliases
DUS3
Chromosome Location
Chr.19: 5785136 - 5791238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020175.2 NP_064560.2 3 - 4 998 89
GenBank mRNA AK023958.1 - 3 - 4 923 89
AK225840.1 - 3 - 4 923 89
AK297037.1 - 3 - 4 783 89
AK315691.1 - 3 - 4 975 89
AL365411.1 - 3 - 4 852 89
BC008362.1 - 2 - 3 792 89
BC009973.1 - 3 - 4 949 89

View Full Product Details