The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184891.1 | NP_001171820.1 | 6 - 7 | 1509 | 70 |
NM_025207.4 | NP_079483.3 | 5 - 6 | 1953 | 70 | |
NM_201398.2 | NP_958800.1 | 6 - 7 | 1509 | 70 | |
GenBank mRNA | AF481877.1 | - | 5 - 6 | 1261 | 70 |
AF520568.1 | - | 5 - 6 | 1874 | 70 | |
AK025979.1 | - | 6 - 7 | 1437 | 70 | |
BC011378.1 | - | 6 - 7 | 1438 | 70 | |
BC020253.1 | - | 4 - 5 | 788 | 70 | |
BC021096.1 | - | 6 - 7 | 1427 | 70 | |
BC032323.1 | - | 6 - 7 | 1444 | 70 | |
DQ458779.1 | - | 5 - 6 | 1552 | 70 | |
U79241.1 | - | 4 - 5 | 1104 | 70 |