Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
Chromosome Location
Chr.X: 112774877 - 112840946 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113490.1 NP_001106962.1 6 - 7 1928 58
NM_133265.2 NP_573572.1 7 - 8 1497 58
XM_005262087.1 XP_005262144.1 9 - 10 2673 58
XM_005262090.1 XP_005262147.1 6 - 7 1170 58
XM_011530875.2 XP_011529177.1 10 - 11 2785 58
XM_017029289.1 XP_016884778.1 8 - 9 1701 58
GenBank mRNA AB028994.3 - 5 - 6 701 58
AF286598.1 - 7 - 8 1497 58
AY987378.1 - 6 - 7 1928 58
BC130294.1 - 5 - 6 918 58

View Full Product Details