The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113490.1 | NP_001106962.1 | 7 - 8 | 2116 | 65 |
NM_133265.2 | NP_573572.1 | 8 - 9 | 1685 | 65 | |
XM_005262087.1 | XP_005262144.1 | 10 - 11 | 2861 | 65 | |
XM_005262090.1 | XP_005262147.1 | 7 - 8 | 1358 | 65 | |
XM_011530875.2 | XP_011529177.1 | 11 - 12 | 2973 | 65 | |
XM_017029289.1 | XP_016884778.1 | 9 - 10 | 1889 | 65 | |
GenBank mRNA | AB028994.3 | - | 6 - 7 | 889 | 65 |
AF286598.1 | - | 8 - 9 | 1685 | 65 | |
AY987378.1 | - | 7 - 8 | 2116 | 65 | |
BC130294.1 | - | 6 - 7 | 1106 | 65 |