Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173909507-173909778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
208077 c.1120G>A p.D374N Chr.1: 173909585-173909585 on Build GRCh38
5735100 c.934A>G p.T312A Chr.1: 173909771-173909771 on Build GRCh38
2150092 c.1119C>T p.V373V Chr.1: 173909586-173909586 on Build GRCh38
5744429 c.1098G>T p.Q366H Chr.1: 173909607-173909607 on Build GRCh38
677071 c.1015T>A p.W339R Chr.1: 173909690-173909690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984249696 Chr.1: 173909739-173909739 on Build GRCh38
rs483352856 Chr.1: 173909689-173909689 on Build GRCh38
rs121909565 Chr.1: 173909564-173909564 on Build GRCh38
rs192187532 Chr.1: 173909700-173909700 on Build GRCh38
rs779331710 Chr.1: 173909535-173909535 on Build GRCh38
rs759734009 Chr.1: 173909753-173909753 on Build GRCh38
rs755808418 Chr.1: 173909683-173909683 on Build GRCh38
rs979120581 Chr.1: 173909722-173909722 on Build GRCh38
rs1010218433 Chr.1: 173909667-173909667 on Build GRCh38
rs775413132 Chr.1: 173909585-173909585 on Build GRCh38
rs934275554 Chr.1: 173909538-173909538 on Build GRCh38
rs5877 Chr.1: 173909724-173909724 on Build GRCh38
rs886045594 Chr.1: 173909651-173909651 on Build GRCh38
rs1035744320 Chr.1: 173909642-173909642 on Build GRCh38
rs777398332 Chr.1: 173909655-173909655 on Build GRCh38
rs909946904 Chr.1: 173909571-173909571 on Build GRCh38
rs959244848 Chr.1: 173909736-173909736 on Build GRCh38
rs774262708 Chr.1: 173909709-173909709 on Build GRCh38
rs565091601 Chr.1: 173909609-173909609 on Build GRCh38
rs758801372 Chr.1: 173909668-173909668 on Build GRCh38
rs770381981 Chr.1: 173909554-173909554 on Build GRCh38
rs199469509 Chr.1: 173909622-173909639 on Build GRCh38
rs762780270 Chr.1: 173909600-173909600 on Build GRCh38
rs776791729 Chr.1: 173909747-173909747 on Build GRCh38
rs758316459 Chr.1: 173909644-173909644 on Build GRCh38
rs766626880 Chr.1: 173909735-173909735 on Build GRCh38
rs780821947 Chr.1: 173909675-173909675 on Build GRCh38
rs774393305 Chr.1: 173909611-173909611 on Build GRCh38
rs483352855 Chr.1: 173909670-173909672 on Build GRCh38
rs745357314 Chr.1: 173909676-173909676 on Build GRCh38
rs749129128 Chr.1: 173909697-173909697 on Build GRCh38
rs1037223864 Chr.1: 173909532-173909532 on Build GRCh38
rs780549243 Chr.1: 173909561-173909561 on Build GRCh38
rs5878 Chr.1: 173909694-173909694 on Build GRCh38
rs767683540 Chr.1: 173909627-173909627 on Build GRCh38
rs369894034 Chr.1: 173909537-173909537 on Build GRCh38
rs121909561 Chr.1: 173909681-173909684 on Build GRCh38
rs753254708 Chr.1: 173909646-173909646 on Build GRCh38
rs762004419 Chr.1: 173909620-173909620 on Build GRCh38
rs761024265 Chr.1: 173909635-173909635 on Build GRCh38
rs773399107 Chr.1: 173909725-173909725 on Build GRCh38
rs376752370 Chr.1: 173909639-173909639 on Build GRCh38
rs200857896 Chr.1: 173909730-173909730 on Build GRCh38
rs745578147 Chr.1: 173909568-173909568 on Build GRCh38
rs121909560 Chr.1: 173909737-173909738 on Build GRCh38
rs567550044 Chr.1: 173909710-173909710 on Build GRCh38
rs765761813 Chr.1: 173909645-173909645 on Build GRCh38
rs149006854 Chr.1: 173909582-173909582 on Build GRCh38
rs551575090 Chr.1: 173909531-173909531 on Build GRCh38
rs373515340 Chr.1: 173909638-173909638 on Build GRCh38
rs960126155 Chr.1: 173909654-173909654 on Build GRCh38
rs773134244 Chr.1: 173909628-173909628 on Build GRCh38
rs863224495 Chr.1: 173909645-173909645 on Build GRCh38

View Full Product Details