Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16227471-16227696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3 XM_017027420.1 XM_017027421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3718135 c.732A>G p.L244L Chr.19: 16227606-16227606 on Build GRCh38
6601558 c.751C>T p.R251C Chr.19: 16227625-16227625 on Build GRCh38
6267430 c.831G>T p.M277I Chr.19: 16227669-16227669 on Build GRCh38
1564640 c.741C>T p.F247F Chr.19: 16227615-16227615 on Build GRCh38
6601560 c.774C>T p.P258P Chr.19: 16227648-16227648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777016904 Chr.19: 16227602-16227602 on Build GRCh38
rs749941507 Chr.19: 16227636-16227636 on Build GRCh38
rs142555840 Chr.19: 16227615-16227615 on Build GRCh38
rs774418578 Chr.19: 16227530-16227530 on Build GRCh38
rs143380976 Chr.19: 16227648-16227648 on Build GRCh38
rs560386574 Chr.19: 16227606-16227606 on Build GRCh38
rs368590185 Chr.19: 16227675-16227675 on Build GRCh38
rs768963580 Chr.19: 16227529-16227529 on Build GRCh38
rs374198455 Chr.19: 16227660-16227660 on Build GRCh38
rs146557925 Chr.19: 16227651-16227651 on Build GRCh38
rs972879096 Chr.19: 16227588-16227588 on Build GRCh38
rs199959764 Chr.19: 16227610-16227610 on Build GRCh38
rs745731808 Chr.19: 16227501-16227501 on Build GRCh38
rs61735380 Chr.19: 16227564-16227564 on Build GRCh38
rs201370085 Chr.19: 16227508-16227508 on Build GRCh38
rs766864704 Chr.19: 16227601-16227601 on Build GRCh38
rs754683740 Chr.19: 16227652-16227652 on Build GRCh38
rs749092235 Chr.19: 16227512-16227512 on Build GRCh38
rs763828773 Chr.19: 16227607-16227607 on Build GRCh38
rs756795435 Chr.19: 16227612-16227612 on Build GRCh38
rs199774903 Chr.19: 16227549-16227549 on Build GRCh38
rs755987949 Chr.19: 16227507-16227507 on Build GRCh38
rs200593906 Chr.19: 16227550-16227550 on Build GRCh38
rs987132870 Chr.19: 16227639-16227639 on Build GRCh38

View Full Product Details