Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13563253-13563502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
UniGene
Hs.504844
Gene Chromosome Location
Chr.12: 13537337-13982012 on Build GRCh38
Transcripts
NM_000834.3 XM_005253351.3 XM_011520628.2 XM_011520629.2 XM_017019219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
430803 c.3896G>A p.R1299H Chr.12: 13563342-13563342 on Build GRCh38
325448 c.3889A>T p.K1297* Chr.12: 13563349-13563349 on Build GRCh38
7370298 c.3981G>T p.K1327N Chr.12: 13563257-13563257 on Build GRCh38
358661 c.3814G>T p.V1272L Chr.12: 13563424-13563424 on Build GRCh38
4040507 c.3957G>A p.P1319P Chr.12: 13563281-13563281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776337201 Chr.12: 13563305-13563305 on Build GRCh38
rs200657400 Chr.12: 13563293-13563293 on Build GRCh38
rs201872316 Chr.12: 13563461-13563461 on Build GRCh38
rs200255226 Chr.12: 13563274-13563274 on Build GRCh38
rs375649732 Chr.12: 13563423-13563424 on Build GRCh38
rs45540034 Chr.12: 13563343-13563343 on Build GRCh38
rs876661100 Chr.12: 13563345-13563345 on Build GRCh38
rs778308238 Chr.12: 13563468-13563468 on Build GRCh38
rs201906121 Chr.12: 13563425-13563425 on Build GRCh38
rs201889168 Chr.12: 13563392-13563392 on Build GRCh38
rs769331485 Chr.12: 13563470-13563470 on Build GRCh38
rs764888979 Chr.12: 13563320-13563320 on Build GRCh38
rs546457168 Chr.12: 13563411-13563411 on Build GRCh38
rs141730031 Chr.12: 13563281-13563281 on Build GRCh38
rs199688138 Chr.12: 13563314-13563314 on Build GRCh38
rs374746622 Chr.12: 13563316-13563316 on Build GRCh38
rs75642143 Chr.12: 13563423-13563423 on Build GRCh38
rs755348109 Chr.12: 13563367-13563367 on Build GRCh38
rs141844705 Chr.12: 13563439-13563439 on Build GRCh38
rs765471479 Chr.12: 13563357-13563359 on Build GRCh38
rs560662030 Chr.12: 13563355-13563355 on Build GRCh38
rs772502958 Chr.12: 13563400-13563400 on Build GRCh38
rs200571186 Chr.12: 13563413-13563413 on Build GRCh38
rs199803550 Chr.12: 13563292-13563292 on Build GRCh38
rs1806200 Chr.12: 13563401-13563401 on Build GRCh38
rs775175973 Chr.12: 13563325-13563325 on Build GRCh38
rs1057398853 Chr.12: 13563426-13563427 on Build GRCh38
rs368885091 Chr.12: 13563383-13563383 on Build GRCh38
rs201947553 Chr.12: 13563441-13563441 on Build GRCh38
rs916745822 Chr.12: 13563348-13563348 on Build GRCh38
rs764172372 Chr.12: 13563332-13563332 on Build GRCh38
rs78765966 Chr.12: 13563431-13563431 on Build GRCh38
rs199935748 Chr.12: 13563442-13563442 on Build GRCh38
rs767709649 Chr.12: 13563342-13563342 on Build GRCh38
rs754079938 Chr.12: 13563440-13563440 on Build GRCh38
rs1017710914 Chr.12: 13563350-13563350 on Build GRCh38
rs774808996 Chr.12: 13563287-13563287 on Build GRCh38
rs748654662 Chr.12: 13563384-13563384 on Build GRCh38
rs750605618 Chr.12: 13563401-13563405 on Build GRCh38
rs760777800 Chr.12: 13563432-13563432 on Build GRCh38
rs372152403 Chr.12: 13563403-13563403 on Build GRCh38
rs564596433 Chr.12: 13563412-13563412 on Build GRCh38
rs757478003 Chr.12: 13563339-13563339 on Build GRCh38
rs779637139 Chr.12: 13563479-13563479 on Build GRCh38
rs201559519 Chr.12: 13563297-13563297 on Build GRCh38
rs796592441 Chr.12: 13563394-13563394 on Build GRCh38
rs753005699 Chr.12: 13563458-13563458 on Build GRCh38
rs201850119 Chr.12: 13563430-13563430 on Build GRCh38
rs531747728 Chr.12: 13563420-13563420 on Build GRCh38
rs376716897 Chr.12: 13563404-13563404 on Build GRCh38
rs946411565 Chr.12: 13563455-13563455 on Build GRCh38
rs964835181 Chr.12: 13563368-13563368 on Build GRCh38
rs762775526 Chr.12: 13563329-13563329 on Build GRCh38
rs200902451 Chr.12: 13563460-13563460 on Build GRCh38
rs372379846 Chr.12: 13563426-13563426 on Build GRCh38
rs143736428 Chr.12: 13563353-13563353 on Build GRCh38
rs150603153 Chr.12: 13563469-13563469 on Build GRCh38
rs749655702 Chr.12: 13563360-13563360 on Build GRCh38

View Full Product Details