Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113204131-113204402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6722587 c.180G>T p.K60N Chr.11: 113204341-113204341 on Build GRCh38
3443427 c.165C>G p.N55K Chr.11: 113204353-113204353 on Build GRCh38
6318515 c.174G>T p.R58R Chr.11: 113204362-113204362 on Build GRCh38
4525723 c.104G>A p.G35E Chr.11: 113204292-113204292 on Build GRCh38
4760229 c.173G>A p.G58E Chr.11: 113204334-113204334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370673346 Chr.11: 113204325-113204325 on Build GRCh38
rs782333170 Chr.11: 113204321-113204321 on Build GRCh38
rs564268734 Chr.11: 113204331-113204331 on Build GRCh38
rs577688149 Chr.11: 113204380-113204380 on Build GRCh38
rs782641265 Chr.11: 113204301-113204301 on Build GRCh38
rs781955531 Chr.11: 113204324-113204324 on Build GRCh38
rs782478693 Chr.11: 113204300-113204300 on Build GRCh38
rs781819959 Chr.11: 113204298-113204298 on Build GRCh38
rs782144440 Chr.11: 113204346-113204346 on Build GRCh38
rs782744343 Chr.11: 113204379-113204379 on Build GRCh38
rs184963226 Chr.11: 113204249-113204249 on Build GRCh38
rs1021111986 Chr.11: 113204153-113204153 on Build GRCh38
rs926101511 Chr.11: 113204225-113204225 on Build GRCh38
rs782177786 Chr.11: 113204316-113204316 on Build GRCh38
rs781816999 Chr.11: 113204258-113204259 on Build GRCh38
rs979335051 Chr.11: 113204205-113204205 on Build GRCh38
rs782770368 Chr.11: 113204274-113204274 on Build GRCh38
rs781923557 Chr.11: 113204361-113204361 on Build GRCh38
rs953635270 Chr.11: 113204234-113204234 on Build GRCh38
rs782679389 Chr.11: 113204314-113204314 on Build GRCh38
rs782402024 Chr.11: 113204312-113204312 on Build GRCh38
rs782073079 Chr.11: 113204243-113204243 on Build GRCh38
rs781981816 Chr.11: 113204189-113204189 on Build GRCh38
rs142223642 Chr.11: 113204224-113204224 on Build GRCh38
rs192623909 Chr.11: 113204174-113204174 on Build GRCh38
rs377478639 Chr.11: 113204291-113204291 on Build GRCh38
rs782583259 Chr.11: 113204296-113204296 on Build GRCh38
rs782251137 Chr.11: 113204305-113204305 on Build GRCh38
rs782375380 Chr.11: 113204330-113204330 on Build GRCh38
rs782115081 Chr.11: 113204254-113204254 on Build GRCh38
rs782820147 Chr.11: 113204357-113204357 on Build GRCh38
rs782080431 Chr.11: 113204364-113204364 on Build GRCh38
rs782751083 Chr.11: 113204248-113204248 on Build GRCh38
rs782555779 Chr.11: 113204293-113204293 on Build GRCh38

View Full Product Details