Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173909733-173909980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899813 c.880C>T p.R294C Chr.1: 173909825-173909825 on Build GRCh38
5735100 c.934A>G p.T312A Chr.1: 173909771-173909771 on Build GRCh38
899814 c.877C>T p.R293W Chr.1: 173909828-173909828 on Build GRCh38
5806885 c.776C>G p.S259* Chr.1: 173909929-173909929 on Build GRCh38
5844545 c.862G>A p.G288S Chr.1: 173909843-173909843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201381904 Chr.1: 173909822-173909822 on Build GRCh38
rs764695432 Chr.1: 173909834-173909834 on Build GRCh38
rs377588972 Chr.1: 173909833-173909833 on Build GRCh38
rs753056696 Chr.1: 173909767-173909767 on Build GRCh38
rs759734009 Chr.1: 173909753-173909753 on Build GRCh38
rs549991084 Chr.1: 173909791-173909791 on Build GRCh38
rs972361914 Chr.1: 173909817-173909817 on Build GRCh38
rs758087836 Chr.1: 173909900-173909900 on Build GRCh38
rs184508490 Chr.1: 173909823-173909823 on Build GRCh38
rs1036017552 Chr.1: 173909838-173909838 on Build GRCh38
rs759002598 Chr.1: 173909769-173909769 on Build GRCh38
rs753390084 Chr.1: 173909871-173909871 on Build GRCh38
rs765632646 Chr.1: 173909764-173909764 on Build GRCh38
rs749956661 Chr.1: 173909778-173909778 on Build GRCh38
rs370190321 Chr.1: 173909835-173909835 on Build GRCh38
rs778650224 Chr.1: 173909872-173909872 on Build GRCh38
rs759821949 Chr.1: 173909830-173909830 on Build GRCh38
rs776662899 Chr.1: 173909828-173909828 on Build GRCh38
rs758329697 Chr.1: 173909889-173909889 on Build GRCh38
rs587776397 Chr.1: 173909824-173909824 on Build GRCh38
rs778915564 Chr.1: 173909820-173909820 on Build GRCh38
rs769991153 Chr.1: 173909831-173909831 on Build GRCh38
rs781308217 Chr.1: 173909947-173909947 on Build GRCh38
rs139275128 Chr.1: 173909897-173909897 on Build GRCh38
rs764455436 Chr.1: 173909770-173909770 on Build GRCh38
rs572313182 Chr.1: 173909827-173909827 on Build GRCh38
rs906122970 Chr.1: 173909931-173909931 on Build GRCh38
rs745879490 Chr.1: 173909883-173909883 on Build GRCh38
rs139463995 Chr.1: 173909847-173909847 on Build GRCh38
rs372820797 Chr.1: 173909819-173909819 on Build GRCh38
rs746961120 Chr.1: 173909928-173909928 on Build GRCh38
rs868179720 Chr.1: 173909942-173909942 on Build GRCh38
rs770852197 Chr.1: 173909939-173909939 on Build GRCh38
rs747142328 Chr.1: 173909825-173909825 on Build GRCh38
rs777786232 Chr.1: 173909921-173909921 on Build GRCh38
rs150507232 Chr.1: 173909795-173909795 on Build GRCh38
rs483352857 Chr.1: 173909851-173909852 on Build GRCh38
rs775845871 Chr.1: 173909832-173909832 on Build GRCh38

View Full Product Details