Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56450486-56450757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620684 c.591A>G p.I197M Chr.20: 56450628-56450628 on Build GRCh38
4581902 c.626C>G p.P209R Chr.20: 56450663-56450663 on Build GRCh38
6160480 c.597C>A p.A199A Chr.20: 56450634-56450634 on Build GRCh38
6052896 c.614G>A p.G205E Chr.20: 56450651-56450651 on Build GRCh38
359711 c.630C>A p.D210E Chr.20: 56450667-56450667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886589948 Chr.20: 56450585-56450585 on Build GRCh38
rs750995912 Chr.20: 56450634-56450634 on Build GRCh38
rs201734849 Chr.20: 56450579-56450579 on Build GRCh38
rs776674677 Chr.20: 56450705-56450705 on Build GRCh38
rs1004096116 Chr.20: 56450661-56450661 on Build GRCh38
rs763896927 Chr.20: 56450548-56450548 on Build GRCh38
rs372643953 Chr.20: 56450707-56450707 on Build GRCh38
rs570282121 Chr.20: 56450564-56450564 on Build GRCh38
rs376902758 Chr.20: 56450718-56450718 on Build GRCh38
rs765282389 Chr.20: 56450708-56450708 on Build GRCh38
rs968070783 Chr.20: 56450538-56450538 on Build GRCh38
rs1031397464 Chr.20: 56450630-56450630 on Build GRCh38
rs953543941 Chr.20: 56450711-56450711 on Build GRCh38
rs750092776 Chr.20: 56450584-56450584 on Build GRCh38
rs774662466 Chr.20: 56450680-56450680 on Build GRCh38
rs745886142 Chr.20: 56450686-56450686 on Build GRCh38
rs770991801 Chr.20: 56450667-56450667 on Build GRCh38
rs569265583 Chr.20: 56450695-56450695 on Build GRCh38
rs966322313 Chr.20: 56450677-56450677 on Build GRCh38
rs754541747 Chr.20: 56450637-56450637 on Build GRCh38
rs193035877 Chr.20: 56450537-56450537 on Build GRCh38
rs188548419 Chr.20: 56450534-56450534 on Build GRCh38
rs868613416 Chr.20: 56450642-56450642 on Build GRCh38
rs766064182 Chr.20: 56450627-56450627 on Build GRCh38
rs6069753 Chr.20: 56450706-56450706 on Build GRCh38
rs558608299 Chr.20: 56450663-56450663 on Build GRCh38
rs758070857 Chr.20: 56450595-56450595 on Build GRCh38
rs748695905 Chr.20: 56450655-56450655 on Build GRCh38
rs1047869688 Chr.20: 56450575-56450575 on Build GRCh38
rs781529808 Chr.20: 56450651-56450651 on Build GRCh38
rs571413631 Chr.20: 56450680-56450681 on Build GRCh38
rs773078792 Chr.20: 56450717-56450717 on Build GRCh38
rs765902094 Chr.20: 56450723-56450723 on Build GRCh38
rs756466731 Chr.20: 56450659-56450659 on Build GRCh38
rs761676974 Chr.20: 56450566-56450566 on Build GRCh38

View Full Product Details