Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46031265-46031532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6434063 c.3274C>T p.H1092Y Chr.17: 46031520-46031520 on Build GRCh38
139063 c.3271C>T p.R1091W Chr.17: 46031523-46031523 on Build GRCh38
980232 c.3306G>A p.P1102P Chr.17: 46031488-46031488 on Build GRCh38
1610370 c.3287C>T p.A1096V Chr.17: 46031507-46031507 on Build GRCh38
6907433 c.3312_3313delCA p.H1104fs*3 Chr.17: 46031481-46031482 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs76025910 Chr.17: 46031335-46031335 on Build GRCh38
rs752732089 Chr.17: 46031388-46031388 on Build GRCh38
rs774194245 Chr.17: 46031474-46031474 on Build GRCh38
rs901807508 Chr.17: 46031472-46031472 on Build GRCh38
rs938927477 Chr.17: 46031286-46031286 on Build GRCh38
rs769590171 Chr.17: 46031467-46031467 on Build GRCh38
rs753191139 Chr.17: 46031495-46031495 on Build GRCh38
rs778296615 Chr.17: 46031431-46031431 on Build GRCh38
rs553624358 Chr.17: 46031429-46031429 on Build GRCh38
rs905971553 Chr.17: 46031325-46031326 on Build GRCh38
rs1043475663 Chr.17: 46031342-46031342 on Build GRCh38
rs778823729 Chr.17: 46031331-46031331 on Build GRCh38
rs943068318 Chr.17: 46031413-46031413 on Build GRCh38
rs567243937 Chr.17: 46031459-46031459 on Build GRCh38
rs1020982547 Chr.17: 46031416-46031416 on Build GRCh38
rs775460035 Chr.17: 46031468-46031468 on Build GRCh38
rs767896466 Chr.17: 46031441-46031444 on Build GRCh38
rs796052598 Chr.17: 46031496-46031496 on Build GRCh38
rs1035821925 Chr.17: 46031326-46031327 on Build GRCh38
rs560719955 Chr.17: 46031499-46031499 on Build GRCh38
rs913401170 Chr.17: 46031309-46031309 on Build GRCh38
rs529738082 Chr.17: 46031507-46031507 on Build GRCh38
rs1001626132 Chr.17: 46031326-46031326 on Build GRCh38
rs947552634 Chr.17: 46031314-46031314 on Build GRCh38
rs765828427 Chr.17: 46031494-46031494 on Build GRCh38
rs373668834 Chr.17: 46031439-46031442 on Build GRCh38
rs747161928 Chr.17: 46031446-46031446 on Build GRCh38
rs754855855 Chr.17: 46031293-46031293 on Build GRCh38
rs372125138 Chr.17: 46031473-46031473 on Build GRCh38
rs143746890 Chr.17: 46031488-46031488 on Build GRCh38
rs989036321 Chr.17: 46031294-46031294 on Build GRCh38
rs776352811 Chr.17: 46031458-46031459 on Build GRCh38
rs761370843 Chr.17: 46031481-46031482 on Build GRCh38
rs546370187 Chr.17: 46031362-46031362 on Build GRCh38
rs78288850 Chr.17: 46031326-46031326 on Build GRCh38
rs542543248 Chr.17: 46031489-46031489 on Build GRCh38
rs746012943 Chr.17: 46031461-46031461 on Build GRCh38
rs768138439 Chr.17: 46031446-46031449 on Build GRCh38
rs764865498 Chr.17: 46031504-46031506 on Build GRCh38

View Full Product Details