Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58696652-58696925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51C
Entrez Gene ID
5889
Gene Name
RAD51 paralog C
Gene Aliases
BROVCA3, FANCO, R51H3, RAD51L2
UniGene
Hs.412587
Gene Chromosome Location
Chr.17: 58692538-58734342 on Build GRCh38
Transcripts
NM_058216.2 NR_103872.1 XM_006722001.3 XM_006722002.3 XM_006722004.3 XM_006722005.3 XM_011525092.2 XM_011525093.2 XM_011525094.2 XM_017024914.1 XM_017024915.1 XM_017024916.1 XM_017024917.1 XM_017024918.1 XM_017024919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6910260 c.487A>C p.S163R Chr.17: 58696775-58696775 on Build GRCh38
4988152 c.454G>A p.A152T Chr.17: 58696742-58696742 on Build GRCh38
6916585 c.463G>A p.A155T Chr.17: 58696751-58696751 on Build GRCh38
1610546 c.459T>G p.G153G Chr.17: 58696747-58696747 on Build GRCh38
2157305 c.461_462delAA p.E154fs*5 Chr.17: 58696749-58696750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767247010 Chr.17: 58696707-58696707 on Build GRCh38
rs779932777 Chr.17: 58696856-58696856 on Build GRCh38
rs776730538 Chr.17: 58696868-58696868 on Build GRCh38
rs770815835 Chr.17: 58696690-58696690 on Build GRCh38
rs876660817 Chr.17: 58696726-58696726 on Build GRCh38
rs587780257 Chr.17: 58696863-58696863 on Build GRCh38
rs111939093 Chr.17: 58696887-58696887 on Build GRCh38
rs745824401 Chr.17: 58696846-58696846 on Build GRCh38
rs587780838 Chr.17: 58696811-58696811 on Build GRCh38
rs730881934 Chr.17: 58696737-58696737 on Build GRCh38
rs864622278 Chr.17: 58696808-58696808 on Build GRCh38
rs537272666 Chr.17: 58696693-58696693 on Build GRCh38
rs577697136 Chr.17: 58696879-58696879 on Build GRCh38
rs745755962 Chr.17: 58696757-58696757 on Build GRCh38
rs774137305 Chr.17: 58696877-58696877 on Build GRCh38
rs776534031 Chr.17: 58696841-58696842 on Build GRCh38
rs140279158 Chr.17: 58696801-58696801 on Build GRCh38
rs375007340 Chr.17: 58696874-58696874 on Build GRCh38
rs145779113 Chr.17: 58696864-58696864 on Build GRCh38
rs376494695 Chr.17: 58696710-58696710 on Build GRCh38
rs587782036 Chr.17: 58696692-58696692 on Build GRCh38
rs28363307 Chr.17: 58696719-58696719 on Build GRCh38
rs746993675 Chr.17: 58696785-58696785 on Build GRCh38
rs768793789 Chr.17: 58696811-58696812 on Build GRCh38
rs761168518 Chr.17: 58696840-58696840 on Build GRCh38
rs553112850 Chr.17: 58696736-58696736 on Build GRCh38
rs765300465 Chr.17: 58696785-58696785 on Build GRCh38
rs753912045 Chr.17: 58696739-58696739 on Build GRCh38
rs587780837 Chr.17: 58696709-58696709 on Build GRCh38
rs776904180 Chr.17: 58696775-58696775 on Build GRCh38
rs770464788 Chr.17: 58696754-58696754 on Build GRCh38
rs763093967 Chr.17: 58696880-58696880 on Build GRCh38
rs876658264 Chr.17: 58696852-58696852 on Build GRCh38
rs765730332 Chr.17: 58696746-58696746 on Build GRCh38
rs1057517812 Chr.17: 58696849-58696850 on Build GRCh38
rs370506962 Chr.17: 58696874-58696875 on Build GRCh38
rs35151472 Chr.17: 58696773-58696773 on Build GRCh38
rs771888635 Chr.17: 58696760-58696760 on Build GRCh38
rs587780256 Chr.17: 58696794-58696794 on Build GRCh38
rs587781680 Chr.17: 58696851-58696851 on Build GRCh38
rs143941948 Chr.17: 58696754-58696755 on Build GRCh38
rs555235745 Chr.17: 58696723-58696723 on Build GRCh38
rs587780255 Chr.17: 58696716-58696716 on Build GRCh38
rs772136509 Chr.17: 58696756-58696769 on Build GRCh38
rs758847241 Chr.17: 58696749-58696749 on Build GRCh38
rs876659719 Chr.17: 58696777-58696777 on Build GRCh38
rs777452778 Chr.17: 58696752-58696752 on Build GRCh38
rs1024076621 Chr.17: 58696866-58696866 on Build GRCh38
rs777351392 Chr.17: 58696831-58696831 on Build GRCh38
rs864622580 Chr.17: 58696731-58696731 on Build GRCh38
rs775213492 Chr.17: 58696763-58696763 on Build GRCh38
rs730881938 Chr.17: 58696706-58696706 on Build GRCh38
rs780353998 Chr.17: 58696773-58696774 on Build GRCh38
rs372385738 Chr.17: 58696825-58696825 on Build GRCh38
rs863224436 Chr.17: 58696705-58696705 on Build GRCh38
rs756727559 Chr.17: 58696836-58696836 on Build GRCh38
rs746870368 Chr.17: 58696860-58696860 on Build GRCh38
rs778731025 Chr.17: 58696756-58696756 on Build GRCh38
rs267606999 Chr.17: 58696702-58696702 on Build GRCh38
rs141621051 Chr.17: 58696875-58696875 on Build GRCh38
rs587780836 Chr.17: 58696696-58696696 on Build GRCh38
rs587780254 Chr.17: 58696694-58696694 on Build GRCh38
rs768903850 Chr.17: 58696769-58696769 on Build GRCh38
rs764177838 Chr.17: 58696728-58696728 on Build GRCh38
rs755879664 Chr.17: 58696751-58696751 on Build GRCh38
rs587781490 Chr.17: 58696790-58696790 on Build GRCh38
rs753475114 Chr.17: 58696835-58696835 on Build GRCh38
rs45553636 Chr.17: 58696741-58696741 on Build GRCh38
rs769486350 Chr.17: 58696747-58696747 on Build GRCh38
rs876660732 Chr.17: 58696802-58696802 on Build GRCh38
rs587782699 Chr.17: 58696790-58696791 on Build GRCh38
rs748811733 Chr.17: 58696683-58696683 on Build GRCh38
rs767061748 Chr.17: 58696891-58696891 on Build GRCh38
rs747825755 Chr.17: 58696750-58696750 on Build GRCh38
rs760436864 Chr.17: 58696765-58696765 on Build GRCh38
rs786201828 Chr.17: 58696817-58696817 on Build GRCh38
rs573992101 Chr.17: 58696780-58696780 on Build GRCh38

View Full Product Details