Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100402417-100402690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7141099 c.1436G>T p.R479L Chr.7: 100402551-100402551 on Build GRCh38
4742390 c.1435C>T p.R479* Chr.7: 100402552-100402552 on Build GRCh38
3394371 c.1471+5G>T p.? Chr.7: 100402511-100402511 on Build GRCh38
3630730 c.1429C>T p.R477C Chr.7: 100402558-100402558 on Build GRCh38
3684369 c.1428T>G p.I476M Chr.7: 100402559-100402559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766317643 Chr.7: 100402557-100402557 on Build GRCh38
rs756471080 Chr.7: 100402561-100402561 on Build GRCh38
rs777789711 Chr.7: 100402565-100402565 on Build GRCh38
rs867660952 Chr.7: 100402516-100402516 on Build GRCh38
rs35111986 Chr.7: 100402651-100402651 on Build GRCh38
rs371378643 Chr.7: 100402572-100402572 on Build GRCh38
rs780458683 Chr.7: 100402617-100402617 on Build GRCh38
rs1046667014 Chr.7: 100402540-100402540 on Build GRCh38
rs777966745 Chr.7: 100402466-100402466 on Build GRCh38
rs981745494 Chr.7: 100402618-100402618 on Build GRCh38
rs762318033 Chr.7: 100402592-100402593 on Build GRCh38
rs28666818 Chr.7: 100402454-100402454 on Build GRCh38
rs557010310 Chr.7: 100402586-100402586 on Build GRCh38
rs375737733 Chr.7: 100402527-100402527 on Build GRCh38
rs753963318 Chr.7: 100402569-100402569 on Build GRCh38
rs200136153 Chr.7: 100402484-100402484 on Build GRCh38
rs767800874 Chr.7: 100402622-100402625 on Build GRCh38
rs779502178 Chr.7: 100402574-100402574 on Build GRCh38
rs377708956 Chr.7: 100402552-100402552 on Build GRCh38
rs768362697 Chr.7: 100402621-100402621 on Build GRCh38
rs765235035 Chr.7: 100402549-100402549 on Build GRCh38
rs558283367 Chr.7: 100402463-100402463 on Build GRCh38
rs200876705 Chr.7: 100402538-100402538 on Build GRCh38
rs541595434 Chr.7: 100402558-100402558 on Build GRCh38
rs368778865 Chr.7: 100402536-100402536 on Build GRCh38
rs906250515 Chr.7: 100402613-100402613 on Build GRCh38
rs373450388 Chr.7: 100402551-100402551 on Build GRCh38
rs774660658 Chr.7: 100402473-100402473 on Build GRCh38
rs776092184 Chr.7: 100402623-100402623 on Build GRCh38
rs973326568 Chr.7: 100402459-100402459 on Build GRCh38
rs772541711 Chr.7: 100402609-100402609 on Build GRCh38
rs202094370 Chr.7: 100402517-100402517 on Build GRCh38
rs771472340 Chr.7: 100402469-100402469 on Build GRCh38
rs1000070255 Chr.7: 100402528-100402528 on Build GRCh38
rs772383063 Chr.7: 100402506-100402506 on Build GRCh38
rs192350517 Chr.7: 100402619-100402619 on Build GRCh38

View Full Product Details