Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113424296-113424545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
UniGene
Hs.73893
Gene Chromosome Location
Chr.11: 113409595-113475279 on Build GRCh38
Transcripts
NM_000795.3 NM_016574.3 XM_017017296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250697 c.136G>A p.A46T Chr.11: 113424516-113424516 on Build GRCh38
5780365 c.277T>G p.Y93D Chr.11: 113424375-113424375 on Build GRCh38
2103333 c.175G>A p.V59M Chr.11: 113424477-113424477 on Build GRCh38
4867801 c.113C>T p.A38V Chr.11: 113424539-113424539 on Build GRCh38
5779974 c.220G>A p.V74I Chr.11: 113424432-113424432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4986918 Chr.11: 113424520-113424520 on Build GRCh38
rs199961459 Chr.11: 113424402-113424402 on Build GRCh38
rs758449566 Chr.11: 113424448-113424448 on Build GRCh38
rs954097445 Chr.11: 113424474-113424474 on Build GRCh38
rs763519955 Chr.11: 113424397-113424397 on Build GRCh38
rs200084134 Chr.11: 113424424-113424424 on Build GRCh38
rs778215584 Chr.11: 113424326-113424326 on Build GRCh38
rs80030508 Chr.11: 113424499-113424499 on Build GRCh38
rs199841276 Chr.11: 113424352-113424352 on Build GRCh38
rs868401982 Chr.11: 113424468-113424468 on Build GRCh38
rs970838089 Chr.11: 113424370-113424370 on Build GRCh38
rs200010436 Chr.11: 113424495-113424495 on Build GRCh38
rs756819930 Chr.11: 113424318-113424318 on Build GRCh38
rs769905247 Chr.11: 113424483-113424483 on Build GRCh38
rs377230925 Chr.11: 113424352-113424352 on Build GRCh38
rs764292347 Chr.11: 113424317-113424317 on Build GRCh38
rs758392774 Chr.11: 113424332-113424332 on Build GRCh38
rs767660986 Chr.11: 113424502-113424502 on Build GRCh38
rs201114741 Chr.11: 113424496-113424496 on Build GRCh38
rs749350345 Chr.11: 113424490-113424490 on Build GRCh38
rs201945675 Chr.11: 113424469-113424469 on Build GRCh38
rs376236416 Chr.11: 113424487-113424487 on Build GRCh38
rs907486029 Chr.11: 113424363-113424363 on Build GRCh38
rs780059943 Chr.11: 113424346-113424346 on Build GRCh38
rs753530257 Chr.11: 113424423-113424423 on Build GRCh38
rs748462272 Chr.11: 113424470-113424470 on Build GRCh38
rs202172997 Chr.11: 113424421-113424421 on Build GRCh38
rs201057025 Chr.11: 113424453-113424453 on Build GRCh38
rs781498037 Chr.11: 113424356-113424356 on Build GRCh38
rs748252813 Chr.11: 113424359-113424359 on Build GRCh38
rs746767289 Chr.11: 113424350-113424350 on Build GRCh38
rs759920076 Chr.11: 113424415-113424415 on Build GRCh38
rs374010633 Chr.11: 113424516-113424516 on Build GRCh38
rs749894030 Chr.11: 113424436-113424436 on Build GRCh38
rs201736234 Chr.11: 113424403-113424403 on Build GRCh38
rs75275037 Chr.11: 113424508-113424508 on Build GRCh38
rs774749525 Chr.11: 113424406-113424406 on Build GRCh38
rs1053323059 Chr.11: 113424432-113424432 on Build GRCh38
rs201795635 Chr.11: 113424517-113424517 on Build GRCh38
rs1038432691 Chr.11: 113424315-113424315 on Build GRCh38
rs866532489 Chr.11: 113424380-113424380 on Build GRCh38
rs201137518 Chr.11: 113424461-113424461 on Build GRCh38
rs750385510 Chr.11: 113424329-113424329 on Build GRCh38
rs76985854 Chr.11: 113424360-113424360 on Build GRCh38
rs751823662 Chr.11: 113424475-113424475 on Build GRCh38
rs769048422 Chr.11: 113424353-113424353 on Build GRCh38
rs139923738 Chr.11: 113424507-113424507 on Build GRCh38
rs201061220 Chr.11: 113424515-113424515 on Build GRCh38
rs202051438 Chr.11: 113424319-113424319 on Build GRCh38
rs762573963 Chr.11: 113424524-113424524 on Build GRCh38
rs773154659 Chr.11: 113424381-113424381 on Build GRCh38
rs764858158 Chr.11: 113424433-113424433 on Build GRCh38

View Full Product Details