Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18782926-18783189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140417 c.2203G>C p.A735P Chr.19: 18783078-18783078 on Build GRCh38
1248962 c.2164C>G p.L722V Chr.19: 18783117-18783117 on Build GRCh38
993156 c.2151G>A p.M717I Chr.19: 18783130-18783130 on Build GRCh38
4075738 c.2122G>A p.D708N Chr.19: 18783159-18783159 on Build GRCh38
4656535 c.2213G>A p.R738H Chr.19: 18783068-18783068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758558539 Chr.19: 18783011-18783011 on Build GRCh38
rs28494505 Chr.19: 18783001-18783001 on Build GRCh38
rs755508135 Chr.19: 18782954-18782954 on Build GRCh38
rs372951104 Chr.19: 18783111-18783111 on Build GRCh38
rs747085396 Chr.19: 18783118-18783118 on Build GRCh38
rs769440522 Chr.19: 18782964-18782964 on Build GRCh38
rs745779375 Chr.19: 18783031-18783031 on Build GRCh38
rs772102125 Chr.19: 18782966-18782971 on Build GRCh38
rs571060152 Chr.19: 18783017-18783017 on Build GRCh38
rs142160846 Chr.19: 18783160-18783160 on Build GRCh38
rs773943832 Chr.19: 18782997-18782997 on Build GRCh38
rs200438524 Chr.19: 18783035-18783035 on Build GRCh38
rs771292272 Chr.19: 18783159-18783159 on Build GRCh38
rs374459827 Chr.19: 18783102-18783102 on Build GRCh38
rs137852655 Chr.19: 18783125-18783125 on Build GRCh38
rs766640350 Chr.19: 18783015-18783015 on Build GRCh38
rs768362782 Chr.19: 18782983-18782983 on Build GRCh38
rs772781703 Chr.19: 18783150-18783150 on Build GRCh38
rs768146721 Chr.19: 18783037-18783037 on Build GRCh38
rs766913050 Chr.19: 18783068-18783068 on Build GRCh38
rs556452100 Chr.19: 18783151-18783151 on Build GRCh38
rs312262904 Chr.19: 18783126-18783126 on Build GRCh38
rs756146893 Chr.19: 18783032-18783032 on Build GRCh38
rs940822896 Chr.19: 18783103-18783103 on Build GRCh38
rs780066431 Chr.19: 18783127-18783127 on Build GRCh38
rs28936368 Chr.19: 18783129-18783129 on Build GRCh38
rs369762722 Chr.19: 18783047-18783047 on Build GRCh38
rs1003270019 Chr.19: 18782958-18782958 on Build GRCh38
rs771687646 Chr.19: 18783048-18783048 on Build GRCh38
rs778901691 Chr.19: 18783135-18783135 on Build GRCh38
rs761432619 Chr.19: 18782998-18782998 on Build GRCh38
rs982663804 Chr.19: 18783051-18783051 on Build GRCh38
rs765456711 Chr.19: 18783023-18783023 on Build GRCh38
rs868294159 Chr.19: 18783161-18783161 on Build GRCh38
rs747790659 Chr.19: 18783147-18783147 on Build GRCh38
rs138174876 Chr.19: 18783062-18783062 on Build GRCh38
rs869320730 Chr.19: 18783057-18783058 on Build GRCh38
rs949107129 Chr.19: 18783021-18783021 on Build GRCh38
rs752882695 Chr.19: 18783095-18783095 on Build GRCh38
rs776802201 Chr.19: 18783069-18783069 on Build GRCh38
rs779485237 Chr.19: 18782974-18782974 on Build GRCh38
rs781024416 Chr.19: 18783025-18783025 on Build GRCh38
rs761081987 Chr.19: 18783063-18783063 on Build GRCh38
rs751668619 Chr.19: 18783119-18783119 on Build GRCh38
rs975316335 Chr.19: 18783041-18783041 on Build GRCh38
rs755950559 Chr.19: 18783120-18783120 on Build GRCh38
rs759998880 Chr.19: 18783167-18783167 on Build GRCh38
rs758502189 Chr.19: 18783026-18783026 on Build GRCh38
rs149551600 Chr.19: 18783128-18783128 on Build GRCh38
rs765401313 Chr.19: 18783077-18783077 on Build GRCh38
rs139001378 Chr.19: 18783148-18783148 on Build GRCh38
rs762512704 Chr.19: 18782981-18782981 on Build GRCh38
rs755087551 Chr.19: 18783022-18783022 on Build GRCh38
rs116499541 Chr.19: 18783046-18783046 on Build GRCh38
rs371523847 Chr.19: 18783036-18783036 on Build GRCh38
rs146075881 Chr.19: 18782980-18782980 on Build GRCh38
rs745619890 Chr.19: 18782957-18782957 on Build GRCh38
rs773825290 Chr.19: 18783003-18783003 on Build GRCh38

View Full Product Details