Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14857817-14858066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs767651966 Chr.X: 14857960-14857961 on Build GRCh38
rs398123537 Chr.X: 14857956-14857957 on Build GRCh38
rs199909156 Chr.X: 14857919-14857919 on Build GRCh38
rs902534750 Chr.X: 14857925-14857925 on Build GRCh38
rs779491229 Chr.X: 14857990-14857991 on Build GRCh38
rs999488499 Chr.X: 14858007-14858007 on Build GRCh38
rs145542355 Chr.X: 14857900-14857900 on Build GRCh38
rs148882589 Chr.X: 14857928-14857928 on Build GRCh38
rs778313516 Chr.X: 14857961-14857961 on Build GRCh38
rs777787636 Chr.X: 14858020-14858023 on Build GRCh38
rs748588943 Chr.X: 14857946-14857946 on Build GRCh38
rs1040489877 Chr.X: 14858032-14858032 on Build GRCh38
rs376945933 Chr.X: 14857911-14857911 on Build GRCh38
rs764156696 Chr.X: 14857880-14857880 on Build GRCh38
rs757202355 Chr.X: 14857899-14857899 on Build GRCh38
rs866138487 Chr.X: 14857941-14857941 on Build GRCh38
rs781251036 Chr.X: 14857939-14857939 on Build GRCh38
rs374256104 Chr.X: 14858008-14858008 on Build GRCh38
rs755514145 Chr.X: 14857987-14857999 on Build GRCh38
rs901963132 Chr.X: 14858033-14858033 on Build GRCh38
rs752699896 Chr.X: 14857957-14857964 on Build GRCh38
rs751649474 Chr.X: 14857910-14857910 on Build GRCh38
rs756433447 Chr.X: 14857952-14857952 on Build GRCh38
rs753812355 Chr.X: 14857882-14857882 on Build GRCh38
rs749814178 Chr.X: 14858000-14858000 on Build GRCh38
rs758565474 Chr.X: 14858006-14858006 on Build GRCh38
rs779894422 Chr.X: 14857957-14857960 on Build GRCh38

View Full Product Details