Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFF
Entrez Gene ID
56947
Gene Name
mitochondrial fission factor
Gene Aliases
C2orf33, GL004
Chromosome Location
Chr.2: 227325151 - 227357836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277061.1 NP_001263990.1 4 - 5 790 66
NM_001277062.1 NP_001263991.1 3 - 4 612 66
NM_001277063.1 NP_001263992.1 3 - 4 612 66
NM_001277064.1 NP_001263993.1 3 - 4 612 66
NM_001277065.1 NP_001263994.1 3 - 4 612 66
NM_001277066.1 NP_001263995.1 2 - 3 500 66
NM_001277067.1 NP_001263996.1 3 - 4 358 66
NM_001277068.1 NP_001263997.1 2 - 3 335 66
NM_020194.5 NP_064579.3 3 - 4 527 66
XM_005246681.3 XP_005246738.1 4 - 5 710 66
XM_005246682.1 XP_005246739.1 4 - 5 710 66
XM_005246683.1 XP_005246740.1 3 - 4 532 66
XM_005246684.1 XP_005246741.1 4 - 5 710 66
XM_005246685.3 XP_005246742.1 4 - 5 710 66
XM_005246686.3 XP_005246743.1 4 - 5 710 66
XM_005246687.3 XP_005246744.1 4 - 5 710 66
XM_005246688.1 XP_005246745.1 4 - 5 710 66
XM_006712637.1 XP_006712700.1 2 - 3 434 66
XM_006712638.1 XP_006712701.1 2 - 3 434 66
XM_006712639.2 XP_006712702.1 2 - 3 434 66
XM_011511500.1 XP_011509802.1 4 - 5 582 66
XM_011511501.1 XP_011509803.1 3 - 4 603 66
XM_017004512.1 XP_016860001.1 4 - 5 774 66
XM_017004513.1 XP_016860002.1 3 - 4 532 66
XM_017004514.1 XP_016860003.1 2 - 3 434 66
XM_017004515.1 XP_016860004.1 2 - 3 434 66
GenBank mRNA AF226049.1 - 2 - 3 255 66
AF246239.1 - 3 - 4 527 66
AF258660.1 - 4 - 5 645 66
AK026137.1 - 3 - 4 513 66
AL517791.3 - 3 - 4 487 66
AL559606.3 - 2 - 3 393 66
BC000797.1 - 2 - 3 284 66
BC093024.1 - 3 - 4 495 66
BI550249.1 - 3 - 4 500 66
BQ653061.1 - 1 - 2 83 66
DA737008.1 - 2 - 3 309 66

View Full Product Details