Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL21A
Entrez Gene ID
151194
Gene Name
methyltransferase like 21A
Gene Aliases
FAM119A, HCA557b, HSPA-KMT
Chromosome Location
Chr.2: 207580631 - 207625928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145280.5 NP_660323.3 1 - 2 136 118
XM_005246338.4 XP_005246395.1 1 - 2 106 118
XM_005246340.3 XP_005246397.1 1 - 2 150 118
XM_017003442.1 XP_016858931.1 1 - 2 117 118
GenBank mRNA AK093812.1 - 1 - 2 136 118
BC033720.1 - 1 - 2 147 118

View Full Product Details