Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAAF1
Entrez Gene ID
123872
Gene Name
dynein axonemal assembly factor 1
Gene Aliases
CILD13, LRRC50, ODA7
Chromosome Location
Chr.16: 84145260 - 84178761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178452.5 NP_848547.4 5 - 6 921 77
XM_006721129.2 XP_006721192.1 5 - 6 921 77
XM_011522853.2 XP_011521155.1 5 - 6 921 77
XM_011522854.2 XP_011521156.1 5 - 6 921 77
XM_011522855.2 XP_011521157.1 5 - 6 921 77
XM_011522857.2 XP_011521159.1 5 - 6 921 77
XM_011522858.2 XP_011521160.1 5 - 6 921 77
XM_017022918.1 XP_016878407.1 5 - 6 921 77
XM_017022919.1 XP_016878408.1 5 - 6 1229 77
GenBank mRNA AK057238.1 - 5 - 6 873 77
AK058059.1 - 1 - 2 178 77
AL833327.1 - 5 - 6 892 77
AL833328.1 - 5 - 6 892 77
AL833336.1 - 5 - 6 892 77
BC024009.2 - 5 - 6 864 77
BX648657.1 - 5 - 6 894 77

View Full Product Details