Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30974836-30975101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRHR
Entrez Gene ID
2692
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
UniGene
Hs.733003
Gene Chromosome Location
Chr.7: 30964021-30979531 on Build GRCh38
Transcripts
NM_000823.3 XM_011515263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3880588 c.858A>G p.K286K Chr.7: 30975016-30975016 on Build GRCh38
5531687 c.836C>T p.S279F Chr.7: 30974994-30974994 on Build GRCh38
1238979 c.851T>C p.I284T Chr.7: 30975009-30975009 on Build GRCh38
5567748 c.875C>T p.S292L Chr.7: 30975033-30975033 on Build GRCh38
1673088 c.877G>A p.V293I Chr.7: 30975035-30975035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958218702 Chr.7: 30974903-30974903 on Build GRCh38
rs538502663 Chr.7: 30974894-30974894 on Build GRCh38
rs374443097 Chr.7: 30975031-30975031 on Build GRCh38
rs776458189 Chr.7: 30975012-30975012 on Build GRCh38
rs769411899 Chr.7: 30975078-30975078 on Build GRCh38
rs200143102 Chr.7: 30974953-30974953 on Build GRCh38
rs113782073 Chr.7: 30974904-30974904 on Build GRCh38
rs1011052399 Chr.7: 30974867-30974867 on Build GRCh38
rs748290640 Chr.7: 30974995-30974995 on Build GRCh38
rs377634222 Chr.7: 30974935-30974935 on Build GRCh38
rs201748357 Chr.7: 30974984-30974984 on Build GRCh38
rs747272834 Chr.7: 30975052-30975052 on Build GRCh38
rs35248685 Chr.7: 30974952-30974952 on Build GRCh38
rs756600187 Chr.7: 30974958-30974958 on Build GRCh38
rs774670785 Chr.7: 30974987-30974987 on Build GRCh38
rs763734599 Chr.7: 30975081-30975081 on Build GRCh38
rs759411734 Chr.7: 30975070-30975070 on Build GRCh38
rs748546851 Chr.7: 30975038-30975038 on Build GRCh38
rs577428185 Chr.7: 30974871-30974871 on Build GRCh38
rs1018841495 Chr.7: 30974875-30974875 on Build GRCh38
rs370074960 Chr.7: 30975034-30975034 on Build GRCh38
rs773436596 Chr.7: 30974996-30974996 on Build GRCh38
rs776671355 Chr.7: 30975068-30975068 on Build GRCh38
rs947708255 Chr.7: 30974917-30974917 on Build GRCh38
rs757801797 Chr.7: 30974947-30974947 on Build GRCh38
rs527387367 Chr.7: 30975033-30975033 on Build GRCh38
rs375674796 Chr.7: 30975037-30975037 on Build GRCh38
rs775551951 Chr.7: 30975080-30975080 on Build GRCh38
rs949902190 Chr.7: 30974954-30974954 on Build GRCh38
rs780559231 Chr.7: 30974968-30974968 on Build GRCh38
rs964210820 Chr.7: 30974890-30974890 on Build GRCh38
rs773069868 Chr.7: 30974992-30974992 on Build GRCh38
rs1029838964 Chr.7: 30975044-30975044 on Build GRCh38
rs770780823 Chr.7: 30975000-30975000 on Build GRCh38
rs559861727 Chr.7: 30975022-30975022 on Build GRCh38
rs4988502 Chr.7: 30974916-30974916 on Build GRCh38
rs999592881 Chr.7: 30975071-30975071 on Build GRCh38
rs34430645 Chr.7: 30975058-30975058 on Build GRCh38
rs981760989 Chr.7: 30974921-30974921 on Build GRCh38
rs147768462 Chr.7: 30974986-30974986 on Build GRCh38
rs772197353 Chr.7: 30975043-30975043 on Build GRCh38
rs759443322 Chr.7: 30975019-30975019 on Build GRCh38
rs141237359 Chr.7: 30974997-30974997 on Build GRCh38
rs369716383 Chr.7: 30975028-30975028 on Build GRCh38
rs777602715 Chr.7: 30974948-30974948 on Build GRCh38
rs200997755 Chr.7: 30975051-30975051 on Build GRCh38

View Full Product Details