The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181783.3 | NP_861448.2 | 3 - 4 | 629 | 103 |
GenBank mRNA | AJ697717.1 | - | 3 - 4 | 499 | 103 |
AK074973.1 | - | 2 - 3 | 256 | 103 | |
AK290436.1 | - | 3 - 4 | 629 | 103 | |
AK310375.1 | - | 3 - 4 | 629 | 103 | |
AK310393.1 | - | 3 - 4 | 629 | 103 | |
BC117175.1 | - | 2 - 3 | 450 | 103 | |
BC117177.1 | - | 2 - 3 | 451 | 103 | |
BX538169.1 | - | 3 - 4 | 602 | 103 | |
CR933632.1 | - | 3 - 4 | 580 | 103 |