Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
Chromosome Location
Chr.12: 88142296 - 88199887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181783.3 NP_861448.2 4 - 5 733 145
XM_011537980.2 XP_011536282.1 3 - 4 508 145
GenBank mRNA AJ697717.1 - 4 - 5 603 145
AK290436.1 - 4 - 5 733 145
AK302755.1 - 4 - 5 733 145
AK310375.1 - 4 - 5 733 145
AK310393.1 - 4 - 5 733 145
BC117175.1 - 3 - 4 554 145
BC117177.1 - 3 - 4 555 145
BX538169.1 - 4 - 5 706 145
CR749244.1 - 4 - 5 685 145
CR749309.1 - 4 - 5 719 145
CR933632.1 - 4 - 5 684 145

View Full Product Details