Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRA8
Entrez Gene ID
346673
Gene Name
stimulated by retinoic acid 8
Gene Aliases
-
Chromosome Location
Chr.7: 135231979 - 135258661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182489.1 NP_872295.1 6 - 7 734 109
XM_011516137.2 XP_011514439.1 6 - 7 1587 109
XM_011516138.2 XP_011514440.1 6 - 7 972 109
XM_011516139.2 XP_011514441.1 7 - 8 1216 109
GenBank mRNA AF513502.1 - 6 - 7 734 109

View Full Product Details