Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23415869-23416126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MHRT
Entrez Gene ID
104564225
Gene Name
myosin heavy chain-associated RNA transcript
Gene Aliases
Myheart
UniGene
Gene Chromosome Location
Chr.14: 23415450-23417595 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6075528 c.4895C>T p.A1632V Chr.14: 23416062-23416062 on Build GRCh38
5043275 c.4899C>A p.N1633K Chr.14: 23416058-23416058 on Build GRCh38
7150973 c.4953+1G>C p.? Chr.14: 23416003-23416003 on Build GRCh38
5882477 c.4953+2T>A p.? Chr.14: 23416002-23416002 on Build GRCh38
1477483 c.4941G>A p.Q1647Q Chr.14: 23416016-23416016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750487401 Chr.14: 23415989-23415989 on Build GRCh38
rs762272480 Chr.14: 23416060-23416060 on Build GRCh38
rs369699564 Chr.14: 23415977-23415977 on Build GRCh38
rs150241539 Chr.14: 23416049-23416049 on Build GRCh38
rs979509908 Chr.14: 23416058-23416058 on Build GRCh38
rs121913648 Chr.14: 23416106-23416108 on Build GRCh38
rs145822086 Chr.14: 23416053-23416053 on Build GRCh38
rs777416671 Chr.14: 23416014-23416014 on Build GRCh38
rs376604168 Chr.14: 23415987-23415987 on Build GRCh38
rs151113658 Chr.14: 23416046-23416046 on Build GRCh38
rs772521188 Chr.14: 23415964-23415964 on Build GRCh38
rs750944331 Chr.14: 23415941-23415941 on Build GRCh38
rs904942736 Chr.14: 23415936-23415936 on Build GRCh38
rs397516232 Chr.14: 23416057-23416057 on Build GRCh38
rs914575620 Chr.14: 23415950-23415950 on Build GRCh38
rs565663412 Chr.14: 23416063-23416063 on Build GRCh38
rs764365579 Chr.14: 23415969-23415969 on Build GRCh38
rs369164804 Chr.14: 23416064-23416064 on Build GRCh38
rs587779393 Chr.14: 23416020-23416020 on Build GRCh38
rs756827282 Chr.14: 23416038-23416038 on Build GRCh38
rs587779415 Chr.14: 23416051-23416051 on Build GRCh38
rs34479915 Chr.14: 23415963-23415964 on Build GRCh38
rs749154313 Chr.14: 23416016-23416016 on Build GRCh38
rs549973937 Chr.14: 23415919-23415919 on Build GRCh38
rs777116428 Chr.14: 23415972-23415972 on Build GRCh38
rs1008225259 Chr.14: 23415999-23415999 on Build GRCh38
rs141122361 Chr.14: 23416048-23416048 on Build GRCh38
rs756142583 Chr.14: 23416008-23416008 on Build GRCh38
rs937244889 Chr.14: 23415938-23415938 on Build GRCh38
rs797045730 Chr.14: 23416071-23416071 on Build GRCh38
rs775990796 Chr.14: 23415965-23415965 on Build GRCh38
rs797044596 Chr.14: 23416105-23416107 on Build GRCh38
rs545875689 Chr.14: 23416056-23416056 on Build GRCh38
rs758571263 Chr.14: 23415990-23415990 on Build GRCh38
rs752682962 Chr.14: 23415993-23415993 on Build GRCh38
rs1044737309 Chr.14: 23415924-23415924 on Build GRCh38
rs769259796 Chr.14: 23416054-23416054 on Build GRCh38
rs751747383 Chr.14: 23416096-23416096 on Build GRCh38
rs745415989 Chr.14: 23415956-23415956 on Build GRCh38
rs45442096 Chr.14: 23416102-23416102 on Build GRCh38
rs955638282 Chr.14: 23415996-23415996 on Build GRCh38
rs766561921 Chr.14: 23416085-23416085 on Build GRCh38
rs397516231 Chr.14: 23416093-23416093 on Build GRCh38
rs864309553 Chr.14: 23416042-23416042 on Build GRCh38
rs373509029 Chr.14: 23415991-23415991 on Build GRCh38
rs981658617 Chr.14: 23415907-23415907 on Build GRCh38
rs765324390 Chr.14: 23416061-23416061 on Build GRCh38
rs761921843 Chr.14: 23415975-23415975 on Build GRCh38
rs777500254 Chr.14: 23415993-23415993 on Build GRCh38
rs933520031 Chr.14: 23416000-23416000 on Build GRCh38
rs371949574 Chr.14: 23415971-23415971 on Build GRCh38
rs749695436 Chr.14: 23416100-23416101 on Build GRCh38
rs778651539 Chr.14: 23416043-23416043 on Build GRCh38
rs1037858653 Chr.14: 23415974-23415974 on Build GRCh38

View Full Product Details