The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153456.3 | NP_703157.2 | 2 - 2 | 7197 | 83 |
GenBank mRNA | AF339796.1 | - | 1 - 1 | 887 | 83 |
AK091343.1 | - | 1 - 1 | 1448 | 83 | |
BC037795.1 | - | 1 - 1 | 1263 | 83 |