Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INAFM1
Entrez Gene ID
255783
Gene Name
InaF motif containing 1
Gene Aliases
PRR24
Chromosome Location
Chr.19: 47274402 - 47275723 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178511.5 NP_848606.3 1 - 1 750 69
NR_037675.1 2 - 2 685 69
GenBank mRNA BC022076.2 - 1 - 1 750 69
BC045749.1 - 1 - 1 750 69
BQ708656.1 - 2 - 2 695 69

View Full Product Details