Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77205144-77205391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1701237 c.220C>T p.L74F Chr.9: 77205345-77205345 on Build GRCh38
4918801 c.198A>G p.K66K Chr.9: 77205323-77205323 on Build GRCh38
1110211 c.253G>T p.E85* Chr.9: 77205378-77205378 on Build GRCh38
5977304 c.242C>A p.A81D Chr.9: 77205367-77205367 on Build GRCh38
4918803 c.198A>G p.K66K Chr.9: 77205323-77205323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745946896 Chr.9: 77205304-77205304 on Build GRCh38
rs773150298 Chr.9: 77205347-77205347 on Build GRCh38
rs991992872 Chr.9: 77205192-77205192 on Build GRCh38
rs111562235 Chr.9: 77205223-77205223 on Build GRCh38
rs778985725 Chr.9: 77205297-77205298 on Build GRCh38
rs115443572 Chr.9: 77205353-77205353 on Build GRCh38
rs983711097 Chr.9: 77205252-77205252 on Build GRCh38
rs1025337167 Chr.9: 77205198-77205198 on Build GRCh38
rs749730342 Chr.9: 77205264-77205264 on Build GRCh38
rs75101662 Chr.9: 77205225-77205226 on Build GRCh38
rs929036133 Chr.9: 77205211-77205211 on Build GRCh38
rs772087325 Chr.9: 77205310-77205310 on Build GRCh38
rs749565313 Chr.9: 77205286-77205287 on Build GRCh38
rs951089005 Chr.9: 77205236-77205236 on Build GRCh38
rs757135329 Chr.9: 77205286-77205288 on Build GRCh38
rs765012017 Chr.9: 77205341-77205341 on Build GRCh38
rs942287737 Chr.9: 77205246-77205246 on Build GRCh38
rs775729702 Chr.9: 77205328-77205328 on Build GRCh38
rs376617644 Chr.9: 77205287-77205287 on Build GRCh38
rs754989468 Chr.9: 77205287-77205288 on Build GRCh38
rs774900899 Chr.9: 77205299-77205299 on Build GRCh38
rs538417363 Chr.9: 77205297-77205297 on Build GRCh38
rs917683770 Chr.9: 77205207-77205207 on Build GRCh38
rs909808978 Chr.9: 77205261-77205261 on Build GRCh38
rs113644204 Chr.9: 77205287-77205287 on Build GRCh38
rs760836129 Chr.9: 77205333-77205333 on Build GRCh38
rs569488756 Chr.9: 77205247-77205247 on Build GRCh38

View Full Product Details