Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68208025-68208276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
UniGene
Hs.584921
Gene Chromosome Location
Chr.15: 68206992-68229742 on Build GRCh38
Transcripts
NM_017882.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361571 c.923G>C p.S308T Chr.15: 68208153-68208153 on Build GRCh38
5655758 c.855T>C p.V285V Chr.15: 68208221-68208221 on Build GRCh38
4056482 c.822G>A p.A274A Chr.15: 68208254-68208254 on Build GRCh38
3503447 c.827G>A p.W276* Chr.15: 68208249-68208249 on Build GRCh38
4056481 c.900G>T p.W300C Chr.15: 68208176-68208176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761969677 Chr.15: 68208071-68208071 on Build GRCh38
rs984903628 Chr.15: 68208087-68208087 on Build GRCh38
rs148949069 Chr.15: 68208236-68208236 on Build GRCh38
rs1043430340 Chr.15: 68208218-68208218 on Build GRCh38
rs774291260 Chr.15: 68208206-68208206 on Build GRCh38
rs747282966 Chr.15: 68208114-68208114 on Build GRCh38
rs374990526 Chr.15: 68208191-68208191 on Build GRCh38
rs977154960 Chr.15: 68208067-68208067 on Build GRCh38
rs752212980 Chr.15: 68208101-68208101 on Build GRCh38
rs776693553 Chr.15: 68208118-68208118 on Build GRCh38
rs35529212 Chr.15: 68208087-68208088 on Build GRCh38
rs751297058 Chr.15: 68208095-68208095 on Build GRCh38
rs754655363 Chr.15: 68208098-68208098 on Build GRCh38
rs57058289 Chr.15: 68208065-68208065 on Build GRCh38
rs962181991 Chr.15: 68208111-68208111 on Build GRCh38
rs746373221 Chr.15: 68208107-68208107 on Build GRCh38
rs753344266 Chr.15: 68208245-68208245 on Build GRCh38
rs779232100 Chr.15: 68208105-68208105 on Build GRCh38
rs373652272 Chr.15: 68208204-68208204 on Build GRCh38
rs755203312 Chr.15: 68208188-68208188 on Build GRCh38
rs774237925 Chr.15: 68208152-68208152 on Build GRCh38
rs768820177 Chr.15: 68208117-68208117 on Build GRCh38
rs758756489 Chr.15: 68208110-68208110 on Build GRCh38
rs567275786 Chr.15: 68208203-68208203 on Build GRCh38
rs154774639 Chr.15: 68208186-68208186 on Build GRCh38
rs528304232 Chr.15: 68208066-68208066 on Build GRCh38
rs759348089 Chr.15: 68208228-68208228 on Build GRCh38
rs1027369458 Chr.15: 68208205-68208205 on Build GRCh38
rs767409056 Chr.15: 68208231-68208231 on Build GRCh38
rs762916725 Chr.15: 68208151-68208151 on Build GRCh38
rs143578698 Chr.15: 68208153-68208153 on Build GRCh38
rs747337816 Chr.15: 68208185-68208185 on Build GRCh38
rs151186473 Chr.15: 68208254-68208254 on Build GRCh38
rs201330867 Chr.15: 68208164-68208164 on Build GRCh38
rs202136006 Chr.15: 68208163-68208163 on Build GRCh38
rs748166231 Chr.15: 68208141-68208141 on Build GRCh38
rs773087251 Chr.15: 68208148-68208148 on Build GRCh38
rs551259455 Chr.15: 68208099-68208099 on Build GRCh38
rs564353601 Chr.15: 68208168-68208168 on Build GRCh38
rs758921701 Chr.15: 68208180-68208180 on Build GRCh38
rs780421624 Chr.15: 68208182-68208182 on Build GRCh38
rs769801040 Chr.15: 68208202-68208202 on Build GRCh38
rs794729220 Chr.15: 68208239-68208247 on Build GRCh38
rs750937323 Chr.15: 68208178-68208178 on Build GRCh38
rs911762608 Chr.15: 68208061-68208061 on Build GRCh38
rs760345410 Chr.15: 68208243-68208243 on Build GRCh38
rs952092082 Chr.15: 68208084-68208084 on Build GRCh38
rs780439427 Chr.15: 68208113-68208113 on Build GRCh38
rs143728911 Chr.15: 68208244-68208244 on Build GRCh38
rs769900670 Chr.15: 68208147-68208147 on Build GRCh38
rs983389507 Chr.15: 68208059-68208059 on Build GRCh38
rs757788122 Chr.15: 68208102-68208102 on Build GRCh38
rs146801142 Chr.15: 68208157-68208157 on Build GRCh38
rs886051446 Chr.15: 68208052-68208052 on Build GRCh38
rs369715206 Chr.15: 68208226-68208226 on Build GRCh38
rs983681712 Chr.15: 68208208-68208208 on Build GRCh38
rs996574482 Chr.15: 68208052-68208052 on Build GRCh38
rs202012876 Chr.15: 68208255-68208255 on Build GRCh38
rs916275768 Chr.15: 68208200-68208200 on Build GRCh38
rs781449103 Chr.15: 68208190-68208190 on Build GRCh38

View Full Product Details