The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321864.1 | NP_001308793.1 | 4 - 5 | 366 | 82 |
NM_001321865.1 | NP_001308794.1 | 4 - 5 | 492 | 82 | |
NM_182575.2 | NP_872381.2 | 5 - 6 | 967 | 82 | |
NR_135832.1 | 4 - 5 | 425 | 82 | ||
XM_005258793.4 | XP_005258850.1 | 5 - 6 | 700 | 82 | |
XM_011526812.2 | XP_011525114.1 | 5 - 6 | 1047 | 82 | |
XM_011526813.2 | XP_011525115.1 | 5 - 6 | 896 | 82 | |
XM_011526815.2 | XP_011525117.1 | 4 - 5 | 1610 | 82 | |
XM_017026646.1 | XP_016882135.1 | 4 - 5 | 1471 | 82 | |
GenBank mRNA | AB195682.1 | - | 5 - 6 | 825 | 82 |
AK301750.1 | - | 4 - 5 | 1610 | 82 | |
AY552609.1 | - | 4 - 5 | 419 | 82 | |
AY552610.1 | - | 4 - 5 | 352 | 82 | |
BC034769.1 | - | 5 - 6 | 967 | 82 |