The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321864.1 | NP_001308793.1 | 6 - 7 | 542 | 65 |
NM_001321865.1 | NP_001308794.1 | 6 - 7 | 705 | 65 | |
NM_182575.2 | NP_872381.2 | 7 - 8 | 1143 | 65 | |
XM_005258793.4 | XP_005258850.1 | 7 - 8 | 876 | 65 | |
XM_011526811.2 | XP_011525113.1 | 5 - 6 | 1566 | 65 | |
XM_011526812.2 | XP_011525114.1 | 7 - 8 | 1223 | 65 | |
XM_011526813.2 | XP_011525115.1 | 7 - 8 | 1072 | 65 | |
XM_011526815.2 | XP_011525117.1 | 6 - 7 | 1786 | 65 | |
XM_017026646.1 | XP_016882135.1 | 6 - 7 | 1647 | 65 | |
GenBank mRNA | AB195682.1 | - | 7 - 8 | 1001 | 65 |
AK301750.1 | - | 6 - 7 | 1786 | 65 | |
AY552609.1 | - | 6 - 7 | 632 | 65 | |
BC034769.1 | - | 7 - 8 | 1143 | 65 |