The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303136.1 | NP_001290065.1 | 1 - 2 | 442 | 53 |
NM_152753.3 | NP_689966.2 | 1 - 2 | 442 | 53 | |
XM_005248943.1 | XP_005249000.1 | 1 - 2 | 457 | 53 | |
XM_005248947.1 | XP_005249004.1 | 1 - 2 | 457 | 53 | |
GenBank mRNA | AF452494.1 | - | 1 - 2 | 91 | 53 |
AK291218.1 | - | 2 - 3 | 163 | 53 | |
AY639608.1 | - | 1 - 2 | 85 | 53 | |
BC052263.1 | - | 1 - 2 | 442 | 53 |