The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256743.1 | NP_001243672.1 | 2 - 3 | 567 | 84 |
GenBank mRNA | BC032004.1 | - | 2 - 3 | 511 | 84 |