The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016059.4 | NP_057143.1 | 1 - 2 | 310 | 151 |
GenBank mRNA | AF151882.1 | - | 1 - 2 | 285 | 151 |
AK026636.1 | - | 1 - 2 | 112 | 151 | |
AY359032.1 | - | 1 - 2 | 287 | 151 | |
BC003048.1 | - | 1 - 2 | 262 | 151 | |
BU195579.1 | - | 1 - 2 | 311 | 151 |