The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032391.2 | NP_115767.1 | 1 - 2 | 193 | 75 |
GenBank mRNA | AF331165.1 | - | 1 - 2 | 193 | 75 |
BC030950.1 | - | 1 - 2 | 191 | 75 | |
BU535394.1 | - | 1 - 2 | 196 | 75 |