Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
Chromosome Location
Chr.12: 42081845 - 42144871 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099650.1 NP_001093120.1 7 - 7 1522 115
NM_173601.1 NP_775872.1 8 - 8 1615 115
XM_017019211.1 XP_016874700.1 8 - 8 1354 115
GenBank mRNA AK125141.1 - 7 - 7 1375 115
AL832209.1 - 4 - 4 752 115
BC015597.2 - 4 - 4 677 115
BC030023.1 - 8 - 8 1623 115
BC039145.1 - 7 - 7 1362 115

View Full Product Details