The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004421.2 | NP_004412.2 | 15 - 15 | 2500 | 157 |
XM_005244731.3 | XP_005244788.1 | 15 - 15 | 2605 | 157 | |
GenBank mRNA | AF006011.1 | - | 15 - 15 | 2456 | 157 |
AK093189.1 | - | 8 - 8 | 1740 | 157 | |
AK095867.1 | - | 8 - 8 | 1718 | 157 | |
BC017225.2 | - | 13 - 13 | 2225 | 157 | |
BC050454.1 | - | 9 - 9 | 1845 | 157 | |
BC111419.1 | - | 1 - 1 | 319 | 157 |