The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006419.2 | NP_006410.1 | 3 - 4 | 277 | 70 |
XM_006714063.3 | XP_006714126.1 | 2 - 3 | 270 | 70 | |
GenBank mRNA | AF029894.1 | - | 2 - 3 | 233 | 70 |
AF044197.1 | - | 3 - 4 | 289 | 70 | |
AJ002211.1 | - | 2 - 3 | 233 | 70 | |
BC012589.1 | - | 2 - 3 | 249 | 70 | |
BI836728.1 | - | 2 - 3 | 229 | 70 | |
CR407624.1 | - | 2 - 3 | 199 | 70 |