Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
-
Chromosome Location
Chr.8: 66493256 - 66518524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152765.3 NP_689978.2 5 - 6 835 79
GenBank mRNA AK055047.1 - 2 - 3 169 79
AK096336.1 - 6 - 7 623 79
AK294261.1 - 4 - 5 423 79
AK308800.1 - 6 - 7 662 79
AK315486.1 - 5 - 6 578 79
BC028400.2 - 5 - 6 559 79

View Full Product Details