Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IGLC1
- Entrez Gene ID
-
3537
- Gene Name
- immunoglobulin lambda constant 1
- Gene Aliases
-
IGLC
- Chromosome Location
-
Chr.22: 22895375 - 22895694 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC012159.1 |
- |
1 - 1 |
60 |
110 |
- Gene Symbol
- IGLL5
- Entrez Gene ID
-
100423062
- Gene Name
- immunoglobulin lambda like polypeptide 5
- Gene Aliases
-
IGL, IGLV, VL-MAR
- Chromosome Location
-
Chr.22: 22887780 - 22895834 on Build GRCh38
View Full Product Details