The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005469.3 | NP_005460.2 | 4 - 5 | 739 | 76 |
XM_005260239.3 | XP_005260296.1 | 3 - 4 | 601 | 76 | |
XM_011528480.2 | XP_011526782.1 | 5 - 6 | 763 | 76 | |
GenBank mRNA | AF014404.1 | - | 4 - 5 | 675 | 76 |
AF124264.1 | - | 4 - 5 | 645 | 76 | |
AK298783.1 | - | 2 - 3 | 339 | 76 | |
BC117155.1 | - | 4 - 5 | 667 | 76 | |
BC117157.1 | - | 4 - 5 | 667 | 76 | |
CR541771.1 | - | 4 - 5 | 645 | 76 | |
CR541778.1 | - | 4 - 5 | 645 | 76 | |
X86032.1 | - | 4 - 5 | 713 | 76 |