The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017754.3 | NP_060224.3 | 1 - 2 | 280 | 85 |
GenBank mRNA | AB126777.1 | - | 1 - 2 | 115 | 85 |
BC137120.1 | - | 1 - 2 | 110 | 85 | |
BC137127.1 | - | 1 - 2 | 110 | 85 | |
DB636320.1 | - | 1 - 2 | 280 | 85 |