The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017754.3 | NP_060224.3 | 8 - 9 | 1265 | 71 |
GenBank mRNA | AB126777.1 | - | 8 - 9 | 1100 | 71 |
BC137120.1 | - | 8 - 9 | 1095 | 71 | |
BC137127.1 | - | 8 - 9 | 1095 | 71 |